Heptanal, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

SIALW254002-1KGEA 118 EUR
SIALW254002-1KG SIALW254002-SAMPLE SIALW254002-8KG SIALW254002-20KG
Heptanal, Sigma-Aldrich®
Heptanal
Formula: C₇H₁₄O
MW: 114,19 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –43 °C
Density: 0,82 g/cm³ (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00007028
UN: 3056
ADR: 3,III
Merck Index: 13,04677

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR