Propionaldehyde, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W292303-1KG-K W292303-20KG-K W292303-4KG-K
SIALW292303-1KG-KEA 112 EUR
SIALW292303-1KG-K SIALW292303-20KG-K SIALW292303-4KG-K
Propionaldehyde, Sigma-Aldrich®
Propionaldehyde
Formula: C₃H₆O
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: –81 °C
Density: 0.811 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 13,07915