1,4-Dichlorobutane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
D59100-1KG D59100-250G
SIALD59100-1KGEA 115 EUR
SIALD59100-1KG SIALD59100-250G
1,4-Dichlorobutane, Sigma-Aldrich®
1,4-Dichlorobutane
Formula: ClH₂C(CH₂)₂CH₂Cl
Boiling Pt: 153.9 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.1457 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001011
CAS Number: 110-56-5
EINECS: 203-778-1
UN: 1993
ADR: 3,III