1-Chloropropane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
C68555-100ML
C68555-500ML
SIALC68555-100MLEA
100
EUR
SIALC68555-100ML
SIALC68555-500ML
1-Chloropropane, Sigma-Aldrich®
1-Chloropropane
Formula:
C₃H₇Cl MW: 78.54 g/mol Boiling Pt: 46 °C (1013 hPa) Melting Pt: –122.8 °C Density: 1.3914 g/cm³ (20 °C) Flash Pt: –31 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000995 CAS Number: 540-54-5 EINECS: 208-749-7 UN: 1278 ADR: 3,II Merck Index: 12,08032 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...