Chloroacetamide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2-Chloroacetamide
C0267-1KG
C0267-100G
C0267-500G
SIALC0267-1KGEA
99.8
EUR
SIALC0267-1KG
SIALC0267-100G
SIALC0267-500G
Chloroacetamide, Sigma-Aldrich®
Chloroacetamide
Formula:
C₂H₄ClNO MW: 93.51 g/mol Boiling Pt: 256 °C (1013 hPa) Melting Pt: 115…117 °C Density: 1.6 g/cm³ (20 °C) Flash Pt: 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008027 CAS Number: 79-07-2 EINECS: 201-174-2 UN: 2811 ADR: 6.1,III Merck Index: 13,02127 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...