4'-Chloroacetanilide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-Chloroacetanilide, Paracetamol impurity J, N-(4-Chlorophenyl)acetamide
M-Clarity™ quality level = MQ100
158631-25G
158631-5G
SIAL158631-25GEA
68.6
EUR
SIAL158631-25G
SIAL158631-5G
4'-Chloroacetanilide, Sigma-Aldrich®
4'-Chloroacetanilide
Formula:
C₈H₈ClNO MW: 169.61 g/mol Boiling Pt: 333 °C (1013 hPa) Melting Pt: 179…180 °C Density: 1.385 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000612 CAS Number: 539-03-7 EINECS: 208-707-8 Merck Index: 13,02128 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...