Trichloroacetic acid (TCA), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: TCA
MQ200
T9159-500G T9159-100G T9159-250G
SIALT9159-500GEA 250 EUR
SIALT9159-500G SIALT9159-100G SIALT9159-250G
Trichloroacetic acid (TCA), Sigma-Aldrich®
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM