3-Chloro-1-iodopropane, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Chloro-3-iodopropane
M-Clarity™ quality level = MQ100
234478-100G 234478-25G
SIAL234478-100GEA 141 EUR
SIAL234478-100G SIAL234478-25G
3-Chloro-1-iodopropane, Sigma-Aldrich®
3-Chloro-1-iodopropane
Formula: C₃H₆ClI
MW: 204.44 g/mol
Boiling Pt: 171 °C (1013 hPa)
Density: 1.904 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001096
CAS Number: 6940-76-7
EINECS: 230-088-8