1,3-Dichloro-2-propanol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
184489-100G 184489-500G
SIAL184489-100GEA 79.1 EUR
SIAL184489-100G SIAL184489-500G
1,3-Dichloro-2-propanol, Sigma-Aldrich®
1,3-Dichloro-2-propanol
Formula: ClH₂CCHOHCH₂Cl
MW: 128.99 g/mol
Boiling Pt: 174
Melting Pt: –4 °C
Density: 1.36 g/cm³ (20 °C)
Flash Pt: 85
Storage Temperature: Refrigerator
MDL Number: MFCD00000951
CAS Number: 96-23-1
EINECS: 202-491-9
UN: 2750
ADR: 6.1 II
Merck Index: 13,03100