4-Chloro-1-butanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Chlorobutan-1-ol
M-Clarity™ quality level = MQ200
278823-250ML 278823-50ML
SIAL278823-250MLEA 451 EUR
SIAL278823-250ML SIAL278823-50ML
4-Chloro-1-butanol, Sigma-Aldrich®
4-Chloro-1-butanol
Formula: C₄H₉ClO
Boiling Pt: 84 °C (1013 hPa)
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002967
CAS Number: 928-51-8
EINECS: 213-175-5
UN: 1993
ADR: 3,III