n-Butironitril, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL538264-100MLEA 78.1 EUR
SIAL538264-100ML SIAL538264-1L
n-Butironitril, Sigma-Aldrich®
n-Butironitril
Formula: C₄H₇N
MW: 69,11 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: –112 °C
Density: 0,79 g/cm³ (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00001968
UN: 2411
ADR: 3,II
Merck Index: 13,01597

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR