Propiononitrile, Sigma-Aldrich®
Supplier: Merck
Synonyms: Propionitrile
M-Clarity™ quality level = MQ200
185590-5ML 185590-100ML 185590-500ML
SIAL185590-5MLEA 39.7 EUR
SIAL185590-5ML SIAL185590-100ML SIAL185590-500ML
Propiononitrile, Sigma-Aldrich®
Propiononitrile
Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 97.2 °C (1013 hPa)
Melting Pt: –93 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00001948
CAS Number: 107-12-0
EINECS: 203-464-4
UN: 2404
ADR: 3,II
Merck Index: 12,08012