Acetonitrile, anhydrous ≥99.8%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
M-Clarity™ quality level = MQ200
271004-100ML 271004-1L 271004-2L 271004-12X100
SIAL271004-100MLEA 93.3 EUR
SIAL271004-100ML SIAL271004-1L SIAL271004-2L SIAL271004-12X100
Acetonitrile, anhydrous ≥99.8%, Sigma-Aldrich®
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM