3-Methoxypropiononitrile, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3-Methoxypropanenitrile, 3-Methoxypropionitrile
65290-1L-F
65290-250ML-F
SIAL65290-1L-FEA
227
EUR
SIAL65290-1L-F
SIAL65290-250ML-F
3-Methoxypropiononitrile, Sigma-Aldrich®
3-Methoxypropiononitrile
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 165 °C (1013 hPa) Melting Pt: –63 °C Density: 0.94 g/cm³ (20 °C) |
MDL Number:
MFCD00001958 CAS Number: 110-67-8 EINECS: 203-790-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...