3-Hydroxypropiononitrile, Sigma-Aldrich®

Supplier: Merck

Synonyms: Ethylene Cyanohydrin, 2-Cyanoethanol, 3-Hydroxypropionitrile, Hydracrylonitrile, β-Hydroxypropiononitrile

MQ200

109924-1KG 109924-250G
SIAL109924-1KGEA 124 EUR
SIAL109924-1KG SIAL109924-250G
3-Hydroxypropiononitrile, Sigma-Aldrich®
3-Hydroxypropiononitrile
Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 228
Melting Pt: –46 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002826
CAS Number: 109-78-4
EINECS: 203-704-8
Merck Index: 13,03828

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR