2-Decanone, Sigma-Aldrich®

Supplier: Merck

Synonyms: Decan-2-one

M-Clarity™ quality level = MQ300

W427100-1KG W427100-SAMPLE
SIALW427100-1KGEA 837 EUR
SIALW427100-1KG SIALW427100-SAMPLE
2-Decanone, Sigma-Aldrich®
2-Decanone
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 3 °C
Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009571
CAS Number: 693-54-9
EINECS: 211-752-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR