2-Heptanone, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ300
W254401-1KG-K
W254401-20KG-K
W254401-8KG-K
SIALW254401-1KG-KEA
115
EUR
SIALW254401-1KG-K
SIALW254401-20KG-K
SIALW254401-8KG-K
2-Heptanone, Sigma-Aldrich®
2-Heptanone
Formula:
C₇H₁₄O MW: 114.19 g/mol Boiling Pt: 150 °C (1013 hPa) Melting Pt: –35 °C Density: 0.8111 g/cm³ (20 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009513 CAS Number: 110-43-0 EINECS: 203-767-1 UN: 1110 ADR: 3,III Merck Index: 13,04683 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...