2-Octanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Octan-2-one
M-Clarity™ quality level = MQ300
W280208-1KG-K W280208-2.5KG- W280208-20KG-K W280208-4KG-K
SIALW280208-1KG-KEA 80.1 EUR
SIALW280208-1KG-K SIALW280208-2.5KG- SIALW280208-20KG-K SIALW280208-4KG-K
2-Octanone, Sigma-Aldrich®
2-Octanone
Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 173
Melting Pt: –16 °C
Density: 0.8171 g/cm³ (20 °C)
Flash Pt: 56
Storage Temperature: Ambient
MDL Number: MFCD00009540
CAS Number: 111-13-7
EINECS: 203-837-1
UN: 1993
ADR: 3 III
Merck Index: 13,04731