3-Heptanone, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
H6511-1L
H6511-250ML
SIALH6511-1LEA
538
EUR
SIALH6511-1L
SIALH6511-250ML
3-Heptanone, Sigma-Aldrich®
3-Heptanone
Formula:
C₇H₁₄O MW: 114.19 g/mol Boiling Pt: 147 °C (1013 hPa) Melting Pt: –37 °C Density: 0.812 g/cm³ (20 °C) Flash Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009483 CAS Number: 106-35-4 EINECS: 203-388-1 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...