3-Hexanone, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W329002-100G W329002-1KG W329002-25G W329002-SAMPLE
SIALW329002-100GEA 125 EUR
SIALW329002-100G SIALW329002-1KG SIALW329002-25G SIALW329002-SAMPLE
3-Hexanone, Sigma-Aldrich®
3-Hexanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 123 °C (1013 hPa)
Melting Pt: –55.4 °C
Density: 0.8216 g/cm³ (20 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009402
CAS Number: 589-38-8
EINECS: 209-645-4
UN: 1224
ADR: 3,II