3-Octanone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W280305-1KG-K W280305-20KG-K W280305-8KG-K
SIALW280305-1KG-KEA 160 EUR
SIALW280305-1KG-K SIALW280305-20KG-K SIALW280305-8KG-K
3-Octanone, Sigma-Aldrich®
3-Octanone
Formula: C₈H₁₆O
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: –18.5 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009515
CAS Number: 106-68-3
EINECS: 203-423-0
UN: 1224
ADR: 3,III
Merck Index: 13,03799