4-Heptanone, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
101745-100ML
101745-500ML
101745-5ML
SIAL101745-100MLEA
63.8
EUR
SIAL101745-100ML
SIAL101745-500ML
SIAL101745-5ML
4-Heptanone, Sigma-Aldrich®
4-Heptanone
Formula:
C₇H₁₄O MW: 114.19 g/mol Boiling Pt: 144 °C (1013 hPa) Melting Pt: –34 °C Density: 0.8116 g/cm³ (20 °C) Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009403 CAS Number: 123-19-3 EINECS: 204-608-9 UN: 2710 ADR: 3,III Merck Index: 13,03379 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...