4-Heptanone, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

W254606-SAMPLE W254606-1KG W254606-4KG W254606-8KG
SIALW254606-SAMPLEEA 54.3 EUR
SIALW254606-SAMPLE SIALW254606-1KG SIALW254606-4KG SIALW254606-8KG
4-Heptanone, Sigma-Aldrich®
4-Heptanone
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 144
Melting Pt: –34 °C
Density: 0.8116 g/cm³ (20 °C)
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00009403
CAS Number: 123-19-3
EINECS: 204-608-9
UN: 2710
ADR: 3 III
Merck Index: 13,03379

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR