Acetoacetanilide 99%
Supplier: Thermo Scientific
Synonyms: 3-Oxo-N-phenyl butamide, 3-Oxo-N-phenyl butanamide
102360010 102365000 102360050
ACRO102360010EA 54.2 EUR
ACRO102360010 ACRO102365000 ACRO102360050
Acetoacetanilide 99%
Acetoacetanilide
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 129 °C (32 mmHg)
Melting Pt: 83…86 °C
Density: 1,26 g/cm³ (20 °C)
Flash Pt: 162 °C
MDL Number: MFCD00008780
CAS Number: 102-01-2
EINECS: 202-996-4
Merck Index: 13,00059
Order Now
Specification Test Results
Appearance White Powder or crystals
Infrared spectrum Conforms
Melting point 83 to 86 °C
HPLC ≥98.5 %