Di-tert-butyl pyrocarbonate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
205249-100G
205249-10G
205249-50G
SIAL205249-100GEA
172
EUR
SIAL205249-100G
SIAL205249-10G
SIAL205249-50G
Di-tert-butyl pyrocarbonate, Sigma-Aldrich®
Di-tert-butyl pyrocarbonate
Formula:
C₁₀H₁₈O₅ Boiling Pt: 235 °C (1013 hPa) Melting Pt: 20…23 °C Density: 1.02 g/cm³ (20 °C) Flash Pt: 37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008805 CAS Number: 24424-99-5 EINECS: 246-240-1 UN: 2930 ADR: 6.1,I |