Di-tert-butyl pyrocarbonate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC
M-Clarity™ quality level = MQ200
361941-25G
361941-5G
SIAL361941-25GEA
236
EUR
SIAL361941-25G
SIAL361941-5G
Di-tert-butyl pyrocarbonate, Sigma-Aldrich®
Di-tert-butyl pyrocarbonate
Formula:
C₁₀H₁₈O₅ MW: 218.25 g/mol Boiling Pt: 235 °C (1013 hPa) Melting Pt: 20…23 °C Density: 1.02 g/cm³ (20 °C) Flash Pt: 37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008805 CAS Number: 24424-99-5 EINECS: 246-240-1 UN: 2930 ADR: 6.1,I |