Diethyl pyrocarbonate
Supplier: Roth Carl
Warning
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
K028.1 K028.3
ROTHK028.1EA 82.3 EUR
ROTHK028.1 ROTHK028.3
Diethyl pyrocarbonate
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088