Diethyl pyrocarbonate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
M-Clarity™ quality level = MQ200
40718-100ML 40718-25ML
SIAL40718-100MLEA 521 EUR
SIAL40718-100ML SIAL40718-25ML
Diethyl pyrocarbonate, Sigma-Aldrich®
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088