Pyruvic acid, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
W297003-1KG-K W297003-250G-K W297003-25KG-K W297003-5KG-K
SIALW297003-1KG-KEA 169 EUR
SIALW297003-1KG-K SIALW297003-250G-K SIALW297003-25KG-K SIALW297003-5KG-K
Pyruvic acid, Sigma-Aldrich®
Pyruvic acid
Formula: C₃H₄O₃
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: 12 °C
Density: 1.58 g/cm³ (20 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205