Sodium pyruvate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
M-Clarity™ quality level = MQ200
P3662-100G P3662-25G
SIALP3662-100GEA 476 EUR
SIALP3662-100G SIALP3662-25G
Sodium pyruvate, Sigma-Aldrich®
Sodium pyruvate
Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4