Oxalacetic acid

Supplier: Molekula

MOLE21309006-25GEA 231 EUR
MOLE21309006-25G
Oxalacetic acid
Oxalacetic acid
Formula: C₄H₄O₅
MW: 132,07 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 182…183 °C
UN: 3261
ADR: 8,II
Merck Index: 12,07041

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR