4,4-Dimethoxybutanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4,4-Dimethoxybutan-2-one, 3-Oxobutyraldehyde dimethyl acetal, Acetoacetaldehyde-1,1-Dimethyl Acetal, Acetylacetaldehyde dimethyl acetal, 4,4-Dimethoxy-2-butanone
M-Clarity™ quality level = MQ100
380067-100ML 380067-500ML
SIAL380067-100MLEA 86.2 EUR
SIAL380067-100ML SIAL380067-500ML
4,4-Dimethoxybutanone, Sigma-Aldrich®
4,4-Dimethoxybutanone
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 178
Melting Pt: –82 °C
Density: 0.9451 g/cm³ (20 °C)
Flash Pt: 58
MDL Number: MFCD00008789
CAS Number: 5436-21-5
EINECS: 226-605-1
UN: 1224
ADR: 3 III