Acetoin, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acetyl methyl carbinol, 3-Hydroxybutanone, 3-Hydroxy-2-butanone

M-Clarity™ quality level = MQ100

W200808-1KG-K W200808-25KG-K W200808-5KG-K
SIALW200808-1KG-KEA 145 EUR
SIALW200808-1KG-K SIALW200808-25KG-K SIALW200808-5KG-K
Acetoin, Sigma-Aldrich®
Acetoin
Formula: C₄H₈O₂
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: 20 °C
Density: 0.998 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00038696
CAS Number: 513-86-0
EINECS: 208-174-1
UN: 2621
ADR: 3,III
Merck Index: 13,00065

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR