Acetoin, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetyl methyl carbinol, 3-Hydroxybutanone, 3-Hydroxy-2-butanone
M-Clarity™ quality level = MQ300
W200832-100G-K W200832-1KG-K W200832-5KG-K
SIALW200832-100G-KEA 91 EUR
SIALW200832-100G-K SIALW200832-1KG-K SIALW200832-5KG-K
Acetoin, Sigma-Aldrich®
Acetoin
Formula: C₄H₈O₂
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: 20 °C
Density: 0.998 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00038696
CAS Number: 513-86-0
EINECS: 208-174-1
UN: 2621
ADR: 3,III
Merck Index: 13,00065