2,3-Hexanedione, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
144169-100G 144169-25G
SIAL144169-100GEA 195 EUR
SIAL144169-100G SIAL144169-25G
2,3-Hexanedione, Sigma-Aldrich®
2,3-Hexanedione
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00009398
CAS Number: 3848-24-6
EINECS: 223-350-8
UN: 1224
ADR: 3,III