2,3-Pentanedione, Sigma-Aldrich®
Supplier: Merck
Synonyms: Pentane-2,3-dione
W284114-SAMPLE W284114-10KG-K W284114-1KG-K
SIALW284114-SAMPLEEA 55.4 EUR
SIALW284114-SAMPLE SIALW284114-10KG-K SIALW284114-1KG-K
2,3-Pentanedione, Sigma-Aldrich®
2,3-Pentanedione
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –52 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009313
CAS Number: 600-14-6
EINECS: 209-984-8
UN: 1224
ADR: 3,II