Acetyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetyl chloride 1M in Dichloromethane
M-Clarity™ quality level = MQ200
00990-100ML 00990-1L 00990-250ML
SIAL00990-100MLEA 51 EUR
SIAL00990-100ML SIAL00990-1L SIAL00990-250ML
Acetyl chloride, Sigma-Aldrich®
Acetyl chloride
Formula: C₂H₃ClO
MW: 78.5 g/mol
Boiling Pt: 50.9 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3,II
Merck Index: 13,00087