Acetyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
114189-1KG 114189-25G 114189-500G
SIAL114189-1KGEA 106 EUR
SIAL114189-1KG SIAL114189-25G SIAL114189-500G
Acetyl chloride, Sigma-Aldrich®
Acetyl chloride
Formula: C₂H₃ClO
Boiling Pt: 50.9 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3,II
Merck Index: 13,00087