Propionyl chloride 98%
Supplier: Thermo Fisher Scientific
Danger
131530250 131532500 131535000 131530025
ACRO131530250EA 25.4 EUR
ACRO131530250 ACRO131532500 ACRO131535000 ACRO131530025
Propionyl chloride 98%
Propionyl chloride
Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 77…79 °C (760 mmHg)
Melting Pt: –94 °C
Density: 1.06 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3 II
Merck Index: 12,08013
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4050 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤20 APHA
Titration RCOCl ≥97.5 %