Propionyl chloride, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

P51559-25G P51559-500G
SIALP51559-25GEA 43.5 EUR
SIALP51559-25G SIALP51559-500G
Propionyl chloride, Sigma-Aldrich®
Propionyl chloride
Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 80
Melting Pt: –94 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 6
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3 II
Merck Index: 12,08013

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR