N,N-Dibutylformamide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Formdibutylamide, Formic acid dibutylamide

M-Clarity™ quality level = MQ200

D46904-100G D46904-25G
SIALD46904-100GEA 79.2 EUR
SIALD46904-100G SIALD46904-25G
N,N-Dibutylformamide, Sigma-Aldrich®
N,N-Dibutylformamide
Formula: C₉H₁₉NO
MW: 157.26 g/mol
Boiling Pt: 240
Melting Pt: –30 °C
Density: 0.8932 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003289
CAS Number: 761-65-9
EINECS: 212-090-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR