N,N-Diethylformamide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Formic acid diethylamide, Formdiethylamide
MQ100
186317-100G 186317-25G
SIAL186317-100GEA 102 EUR
SIAL186317-100G SIAL186317-25G
N,N-Diethylformamide, Sigma-Aldrich®
N,N-Diethylformamide
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 176
Density: 0.908 g/cm³ (20 °C)
Flash Pt: 65
Storage Temperature: Ambient
MDL Number: MFCD00003287
CAS Number: 617-84-5
EINECS: 210-533-2