Acetamide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
00160-1KG 00160-250G 00160-50G
SIAL00160-1KGEA 122 EUR
SIAL00160-1KG SIAL00160-250G SIAL00160-50G
Acetamide, Sigma-Aldrich®
Acetamide
Formula: C₂H₅NO
MW: 59.07 g/mol
Boiling Pt: 221.1
Melting Pt: 78…81 °C
Density: 1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008023
CAS Number: 60-35-5
EINECS: 200-473-5
Merck Index: 13,00044