Acetamide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
695122-1G 695122-5G
SIAL695122-1GEA 24.6 EUR
SIAL695122-1G SIAL695122-5G
Acetamide, Sigma-Aldrich®
Acetamide
Formula: C₂H₅NO
MW: 59.07 g/mol
Boiling Pt: 221.1
Melting Pt: 78…81 °C
Density: 1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008023
CAS Number: 60-35-5
EINECS: 200-473-5
Merck Index: 13,00044