Urea ≥99.3%
Supplier: Thermo Fisher Scientific
Purity: 99.3+%
CAS No.: 57-13-6
Molecular Formula: CH4N2O
Formula Weight: 60.06
Physical Form: Solid Crystalline
Density: 1.335
Appearance: White
Odour: Ammonia-like
pH: 9,2-9,5
Melting Point: 132-135°
Beilstein Reference: 635724
Merck Reference: 14,9867
MDL No.: MFCD00008022
Formula:
NH₂CONH₂ Boiling Pt: 197 °C (1013 hPa) Melting Pt: 132…135 °C Density: 1.335 Storage Temperature: Ambient |
MDL Number:
MFCD00008022 CAS Number: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 14,9867 |