Urea
Supplier: DIAGNOSTIC SYSTEMS (DIASYS)
DIAS131159910026EA 0 EUR
DIAS131159910026
Urea
Urea
Formula: NH₂CONH₂
MW: 60,06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132,7 °C
Density: 1,34 g/cm³ (20 °C)
MDL Number: MFCD00008022
CAS Number: 57-13-6
Merck Index: 13,09935
https://si.vwr.com/stibo/web/std.lang.all/70/46/2297046.jpg