Urea, Sigma-Aldrich®
Supplier: Merck
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
51456-2.5KG 51456-500G
SIAL51456-2.5KGEA 307 EUR
SIAL51456-2.5KG SIAL51456-500G
Urea, Sigma-Aldrich®
Urea
Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM