Acetohydrazide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Acetic hydrazide, Acethydrazide
M-Clarity™ quality level = MQ200
A8309-100G
A8309-25G
A8309-5G
SIALA8309-100GEA
282
EUR
SIALA8309-100G
SIALA8309-25G
SIALA8309-5G
Acetohydrazide, Sigma-Aldrich®
Acetohydrazide
Formula:
C₂H₆N₂O MW: 74.08 g/mol Boiling Pt: 246 °C (1013 hPa) Melting Pt: 63…65 °C Flash Pt: >100 °C |
MDL Number:
MFCD00007610 CAS Number: 1068-57-1 EINECS: 213-948-7 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...