Adipohydrazide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Adipyl hydrazide
M-Clarity™ quality level = MQ300
A0638-100G
A0638-25G
SIALA0638-100GEA
328
EUR
SIALA0638-100G
SIALA0638-25G
Adipohydrazide, Sigma-Aldrich®
Adipohydrazide
Formula:
C₆H₁₄N₄O₂ MW: 174.2 g/mol Boiling Pt: 446 Melting Pt: 181…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007614 CAS Number: 1071-93-8 EINECS: 213-999-5 |