Hydroxyurea, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
M-Clarity™ quality level = MQ200
H8627-100G H8627-10G H8627-1G H8627-25G H8627-5G
SIALH8627-100GEA 1470 EUR
SIALH8627-100G SIALH8627-10G SIALH8627-1G SIALH8627-25G SIALH8627-5G
Hydroxyurea, Sigma-Aldrich®
Hydroxyurea
Formula: CH₄N₂O₂
MW: 76.05 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00007943
CAS Number: 127-07-1
EINECS: 204-821-7
Merck Index: 13,04874